4,6-Dichloro-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

22818-1G 22818-25G 22818-500MG 22818-5G
76911-026EA 231.28 USD
76911-026 76911-028 76911-030 76911-032
4,6-Dichloro-1H-indazole ≥95%
4,6-Dichloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MW: 187.026 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781384
CAS Number: 885519-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR