5-Fluoro-m-cresol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Fluoro-3-hydroxy-5-methylbenzene, 3-Fluoro-5-methylphenol

22805-25G 22805-5G
76911-018EA 187.78 USD
76911-018 76911-020
5-Fluoro-m-cresol ≥97%
5-Fluoro-m-cresol

Precaución: for Research Use Only

Formula: C₇H₇FO
MW: 126.13
Storage Temperature: Ambient
MDL Number: MFCD04115885
CAS Number: 216976-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR