3-Bromo-2-fluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

22798-25G 22798-5G
76911-010EA 595.17 USD
76911-010 76911-012
3-Bromo-2-fluoro-N-methoxy-N-methylbenzamide ≥97%
3-Bromo-2-fluoro-N-methoxy-N-methylbenzamide
Formula: C₉H₉BrFNO₂
MW: 262.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21609714
CAS Number: 680610-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR