3-Bromo-2-iodoquinoline ≥95%

Supplier: AOBChem USA

Warning

22788-1G 22788-25G 22788-5G
76910-996EA 84.88 USD
76910-996 76910-998 76911-000
3-Bromo-2-iodoquinoline ≥95%
3-Bromo-2-iodoquinoline
Formula: C₉H₅BrIN
MW: 333.951 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16621398
CAS Number: 898559-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR