4-Bromo-2,5-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22771-1G 22771-500MG 22771-5G
76910-988EA 530.45 USD
76910-988 76910-990 76910-992
4-Bromo-2,5-dichlorobenzaldehyde ≥97%
4-Bromo-2,5-dichlorobenzaldehyde
Formula: C₇H₃BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD18452868
CAS Number: 1349716-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR