3-Bromo-4,5-difluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

22738-1G 22738-25G 22738-500MG 22738-5G
76910-938EA 142.17 USD
76910-938 76910-940 76910-942 76910-944
3-Bromo-4,5-difluorobenzaldehyde ≥95%
3-Bromo-4,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD22056748
CAS Number: 1143502-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR