3-Bromo-4,5-dichloropyridine ≥95%

Supplier: AOBChem USA

Warning

22688-25G 22688-5G
76910-928EA 492.26 USD
76910-928 76910-930
3-Bromo-4,5-dichloropyridine ≥95%
3-Bromo-4,5-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD17014545
CAS Number: 1001056-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR