(3-(Methoxycarbonyl)-5-methylphenyl)boronic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Methoxycarbonyl-5-methylphenylboronic acid
22651-1G
22651-25G
22651-500MG
22651-5G
76910-918EA
267.35
USD
76910-918
76910-920
76910-922
76910-924
(3-(Methoxycarbonyl)-5-methylphenyl)boronic acid ≥97% research grade
(3-(Methoxycarbonyl)-5-methylphenyl)boronic acid
Formula:
C₉H₁₁BO₄ MW: 193.99 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09751432 CAS Number: 929626-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...