3,5-Dichloro-4-methoxybenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
22639-1G
22639-25G
22639-5G
76910-906EA
62.6
USD
76910-906
76910-908
76910-910
3,5-Dichloro-4-methoxybenzaldehyde ≥97%
3,5-Dichloro-4-methoxybenzaldehyde
Formula:
C₈H₆Cl₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD04070684 CAS Number: 41727-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...