3,5-Dibromo-pyridine-2-carbaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Dibromopicolinaldehyde

22579-1G 22579-25G 22579-5G
76910-878EA 91.24 USD
76910-878 76910-880 76910-882
3,5-Dibromo-pyridine-2-carbaldehyde ≥97% research grade
3,5-Dibromo-pyridine-2-carbaldehyde
Formula: C₆H₃Br₂NO
MW: 264.902 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08704598
CAS Number: 898559-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR