3-Bromo-5-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

22542-1G 22542-25G 22542-500MG 22542-5G
76910-868EA 171.87 USD
76910-868 76910-870 76910-872 76910-874
3-Bromo-5-methoxybenzyl alcohol ≥97%
3-Bromo-5-methoxybenzylalkohol
Formula: C₈H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00229988
CAS Number: 262450-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR