4-(Trifluoromethyl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22518-1G 22518-25G 22518-500MG 22518-5G
76910-844EA 167.63 USD
76910-844 76910-846 76910-848 76910-850
4-(Trifluoromethyl)-1H-indazole ≥97%
4-(Trifluoromethyl)-1H-indazole
Formula: C₈H₅F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD09263211
CAS Number: 1000339-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR