3,5-Difluoro-4-isopropoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

22494-1G 22494-25G 22494-5G
76910-830EA 64.72 USD
76910-830 76910-832 76910-834
3,5-Difluoro-4-isopropoxyphenylboronic acid ≥97%
3,5-Difluoro-4-isopropoxyphenylboronic acid
Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20231442
CAS Number: 1451390-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR