3,5-Difluoro-2-hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

22471-1G 22471-25G 22471-500MG 22471-5G
76910-820EA 245.07 USD
76910-820 76910-822 76910-824 76910-826
3,5-Difluoro-2-hydroxyphenylboronic acid pinacol ester ≥97%
3,5-Difluoro-2-hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₅BF₂O₃
MW: 256.054 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996285
CAS Number: 1451391-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR