3-(Dimethylamino)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(N,N-Dimethylamino)benzaldehyde

22453-1G 22453-25G 22453-5G
76910-806EA 114.58 USD
76910-806 76910-808 76910-810
3-(Dimethylamino)benzaldehyde ≥97%
3-(Dimethylamino)benzaldehyde
Formula: C₉H₁₁NO
MW: 149.1897 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05148649
CAS Number: 619-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR