3-((Dimethylamino)methyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-(N,N-Dimethylaminomethyl)phenylboronic acid

22422-1G 22422-25G 22422-5G
76910-794EA 85.94 USD
76910-794 76910-796 76910-798
3-((Dimethylamino)methyl)phenylboronic acid ≥95%
3-((Dimethylamino)methyl)phenylboronic acid
Formula: C₉H₁₄BNO₂
Storage Temperature: Refrigerator
CAS Number: 819849-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR