(4-Acetamidophenyl)boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide, 4-Acetylaminophenylboronic acid pinacol ester, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide

22372-25G 22372-5G
76910-788EA 346.92 USD
76910-788 76910-790
(4-Acetamidophenyl)boronic acid pinacol ester ≥97%
(4-Acetamidophenyl)boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₃
MW: 261.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093722
CAS Number: 214360-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR