(3-Bromo-2-fluoro-6-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-fluoro-6-methoxyphenylboronic acid

22273-1G 22273-25G 22273-5G
76910-774EA 94.43 USD
76910-774 76910-776 76910-778
(3-Bromo-2-fluoro-6-methoxyphenyl)boronic acid ≥97%
(3-Bromo-2-fluoro-6-methoxyphenyl)boronic acid
Formula: C₇H₇BBrFO₃
MW: 248.84 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026725
CAS Number: 1309980-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR