4,6-Dibromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22116-1G 22116-25G 22116-500MG 22116-5G
76910-730EA 205.82 USD
76910-730 76910-732 76910-734 76910-736
4,6-Dibromo-1H-indazole ≥97%
4,6-Dibromo-1H-indazole
Formula: C₇H₄Br₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD01606236
CAS Number: 447430-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR