3-Bromo-2-chloro-4-methoxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

22042-1G 22042-25G 22042-500MG 22042-5G
76910-720EA 192.03 USD
76910-720 76910-722 76910-724 76910-726
3-Bromo-2-chloro-4-methoxypyridine ≥97% research grade
3-Bromo-2-chloro-4-methoxypyridine
Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD14705097
CAS Number: 144584-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR