3-Bromo-2-methylpyridine-6-carboxamide ≥95%

Supplier: AOBChem USA

Warning

21989-1G 21989-25G 21989-500MG 21989-5G
76910-704EA 165.51 USD
76910-704 76910-706 76910-708 76910-710
3-Bromo-2-methylpyridine-6-carboxamide ≥95%
3-Bromo-2-methylpyridine-6-carboxamide
Formula: C₇H₇BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD19105401
CAS Number: 1228014-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR