3-Propoxy-5-trifluoromethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21959-1G 21959-25G 21959-5G
76910-670EA 64.72 USD
76910-670 76910-672 76910-674
3-Propoxy-5-trifluoromethylphenylboronic acid ≥97%
3-Propoxy-5-trifluoromethylphenylboronic acid
Formula: C₁₀H₁₂BF₃O₃
MW: 248.007 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16660267
CAS Number: 1256345-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR