3-Cyano-5-methylphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Cyano-5-methylphenylboronic acid pinacol ester, 3-Methyl-5-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzonitrile

21895-1G 21895-25G 21895-500MG 21895-5G
76910-634EA 173.99 USD
76910-634 76910-636 76910-638 76910-640
3-Cyano-5-methylphenylboronic acid, pinacol ester ≥97%
3-Cyano-5-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₂
MW: 243.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383798
CAS Number: 1220219-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR