4-(Methylcarbamoyl)-2-methoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

21868-1G 21868-25G 21868-500MG 21868-5G
76910-620EA 379.81 USD
76910-620 76910-622 76910-624 76910-626
4-(Methylcarbamoyl)-2-methoxyphenylboronic acid ≥95%
4-(Methylcarbamoyl)-2-methoxyphenylboronic acid
Formula: C₉H₁₂BNO₄
MW: 209.007 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19105391
CAS Number: 1451391-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR