3-Fluoro-4-pyridone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-hydroxypyridine, 3-Fluoropyridin-4-ol

21834-25G 21834-5G
76910-608EA 630.18 USD
76910-608 76910-610
3-Fluoro-4-pyridone ≥97%
3-Fluoro-4-pyridone

Precaución: for Research Use Only

Formula: C₅H₄FNO
Storage Temperature: Ambient
CAS Number: 22282-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR