3-Chloro-5-methoxyphenol ≥95%
Supplier: AOBChem USA
|
Warning
|
21792-25G
21792-5G
76910-596EA
354.35
USD
76910-596
76910-598
3-Chloro-5-methoxyphenol ≥95%
3-Chloro-5-methoxyphenol
Precaución: for Research Use Only
Formula:
C₇H₇ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00002260 CAS Number: 65262-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...