4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(2,2,2-Trimethylacetamino)pyridine-3-carboxylic acid

21728-1G 21728-25G 21728-5G 21728-500MG
76910-582EA 88.06 USD
76910-582 76910-584 76910-586 77649-294
4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid ≥97%
4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid
Formula: C₁₁H₁₄N₂O₃
MW: 222.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407344
CAS Number: 125867-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR