4-(4-Thiomorpholinylcarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-(Thiomorpholine-4-carbonyl)phenylboronic acid, (4-(Thiomorpholine-4-carbonyl)phenyl)boronic acid

21609-1G 21609-25G 21609-500MG 21609-5G
76910-556EA 171.87 USD
76910-556 76910-558 76910-560 76910-562
4-(4-Thiomorpholinylcarbonyl)phenylboronic acid ≥97% research grade
4-(4-Thiomorpholinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₄BNO₃S
MW: 251.1100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659879
CAS Number: 850568-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR