3-Bromo-5-chloro-4-pyridinecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

21554-1G 21554-25G 21554-5G
76910-542EA 88.06 USD
76910-542 76910-544 76910-546
3-Bromo-5-chloro-4-pyridinecarboxylic acid ≥97%
3-Bromo-5-chloro-4-pyridinecarboxylic acid
Formula: C₆H₃BrClNO₂
MW: 236.451 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185794
CAS Number: 1214377-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR