3,4-Difluorosalicylaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,4-Difluoro-2-hydroxybenzaldehyde

21543-1G 21543-25G 21543-5G
76910-532EA 117.76 USD
76910-532 76910-534 76910-536
3,4-Difluorosalicylaldehyde ≥97%
3,4-Difluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
MW: 158.1023 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275283
CAS Number: 502762-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR