3-Bromo-5-methoxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-methoxy-4-methyl-pyridine, 3-Bromo-5-methoxy-4-picoline

21419-1G 21419-25G 21419-5G
76910-500EA 62.6 USD
76910-500 76910-502 76910-504
3-Bromo-5-methoxy-4-methylpyridine ≥97%
3-Bromo-5-methoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257495
CAS Number: 70201-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR