3-Bromo-o-cresol ≥97%

Supplier: AOBChem USA

Warning

21294-25G 21294-5G
76910-480EA 219.61 USD
76910-480 76910-482
3-Bromo-o-cresol ≥97%
3-Bromo-o-cresol

Precaución: for Research Use Only

Formula: C₇H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD11100990
CAS Number: 7766-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR