4-Bromo-2-chloro-1-propoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

21219-25G 21219-5G
76910-468EA 595.17 USD
76910-468 76910-470
4-Bromo-2-chloro-1-propoxybenzene ≥97% research grade
4-Bromo-2-chloro-1-propoxybenzene
Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907660
CAS Number: 1309933-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR