3-Fluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-(pyrolidine-1-carbonyl)phenylboronic acid, (3-Fluoro-4-(pyrrolidine-1-carbonyl)phenyl)boronic acid

21207-1G 21207-25G 21207-5G
76910-456EA 96.55 USD
76910-456 76910-458 76910-460
3-Fluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥97%
3-Fluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₃BFNO₃
MW: 237.0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08436024
CAS Number: 874289-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR