4-(tert-Butylcarbamoyl)-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(tert-Butylcarbamoyl)-3-fluorophenyl)boronic acid

21201-1G 21201-25G 21201-500MG 21201-5G
76910-442EA 130.5 USD
76910-442 76910-444 76910-446 76910-448
4-(tert-Butylcarbamoyl)-3-fluorophenylboronic acid ≥97%
4-(tert-Butylcarbamoyl)-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08689525
CAS Number: 874289-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR