3-Fluoro-4-(phenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21199-1G 21199-25G 21199-500MG 21199-5G
76910-426EA 130.5 USD
76910-426 76910-428 76910-430 76910-432
3-Fluoro-4-(phenylcarbamoyl)phenylboronic acid ≥97%
3-Fluoro-4-(phenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08436025
CAS Number: 874288-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR