4-(Cyclohexylcarbamoyl)-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Cyclohexylcarbamoyl)-3-fluorophenyl)boronic acid

21195-1G 21195-25G 21195-5G
76910-412EA 96.55 USD
76910-412 76910-414 76910-416
4-(Cyclohexylcarbamoyl)-3-fluorophenylboronic acid ≥97%
4-(Cyclohexylcarbamoyl)-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇BFNO₃
MW: 265.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08436036
CAS Number: 874289-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR