3-Fluoro-4-(methylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, (3-Fluoro-4-(methylcarbamoyl)phenyl)boronic acid

21193-1G 21193-25G 21193-5G
76910-400EA 66.84 USD
76910-400 76910-402 76910-404
3-Fluoro-4-(methylcarbamoyl)phenylboronic acid ≥97%
3-Fluoro-4-(methylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BFNO₃
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436026
CAS Number: 849833-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR