3-Bromo-5-(methylthio)pyridine ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-methylthiopyridine

21147-1G 21147-25G 21147-5G
76910-366EA 76.39 USD
76910-366 76910-368 76910-370
3-Bromo-5-(methylthio)pyridine ≥95% research grade
3-Bromo-5-(methylthio)pyridine
Formula: C₆H₆BrNS
Storage Temperature: Refrigerator
MDL Number: MFCD08689537
CAS Number: 142137-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR