4-(Benzyloxy)-2,6-difluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Benzyloxy-2,6-difluorobenzaldehyde

21094-1G 21094-25G 21094-500MG 21094-5G
76910-350EA 177.18 USD
76910-350 76910-352 76910-354 76910-356
4-(Benzyloxy)-2,6-difluorobenzaldehyde ≥95%
4-(Benzyloxy)-2,6-difluorobenzaldehyde
Formula: C₁₄H₁₀F₂O₂
MW: 248.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072454
CAS Number: 918524-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR