4-Bromo-1-(1-ethoxyethyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-1-(1-ethoxy-ethyl)-1H-pyrazole

21082-25G 21082-5G
76910-340EA 204.76 USD
76910-340 76910-342
4-Bromo-1-(1-ethoxyethyl)-1H-pyrazole ≥97%
4-Bromo-1-(1-ethoxyethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₁BrN₂O
MW: 219.079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543653
CAS Number: 1024120-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR