4-Bromo-2,6-difluoro-α-methylbenzyl alcohol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2,6-difluoro-.alpha.-methyl-benzenemethanol, 1-(4-Bromo-2,6-difluorophenyl)ethanol

20999-25G 20999-5G
76910-332EA 240.83 USD
76910-332 76910-334
4-Bromo-2,6-difluoro-α-methylbenzyl alcohol ≥97% research grade
4-Bromo-2,6-difluoro-α-methylbenzyl alcohol
Formula: C₈H₇BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD25459281
CAS Number: 1214900-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR