Methyl 4-bromo-2-(trifluoromethyl)benzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Bromo-2-(trifluoromethyl)benzoic acid methyl ester
20983-25G
20983-5G
76910-322EA
187.78
USD
76910-322
76910-324
Methyl 4-bromo-2-(trifluoromethyl)benzoate ≥97%
Methyl 4-bromo-2-(trifluoromethyl)benzoate
Precaución: for Research Use Only
Formula:
C₉H₆BrF₃O₂ MW: 283.042 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12761599 CAS Number: 957207-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...