3-Chloro-2-formyl-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

20761-1G 20761-25G 20761-500MG 20761-5G
76910-302EA 233.4 USD
76910-302 76910-304 76910-306 76910-308
3-Chloro-2-formyl-4-methylpyridine ≥97%
3-Chloro-2-formyl-4-methylpyridine
Formula: C₇H₆ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD08277284
CAS Number: 884495-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR