3-(Morpholinomethyl)phenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-(Morpholinomethyl)phenyl)boronic acid

20697-1G 20697-25G 20697-5G
76910-282EA 88.06 USD
76910-282 76910-284 76910-286
3-(Morpholinomethyl)phenylboronic acid ≥95% research grade
3-(Morpholinomethyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08060623
CAS Number: 397843-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR