4-Bromo-2-(trifluoromethyl)benzeneboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Bromo-2-(trifluoromethyl)phenylboronic acid, 2-Borono-5-bromobenzotrifluoride, (4-Bromo-2-(trifluoromethyl)phenyl)boronic acid
20657-1G
20657-25G
20657-5G
76910-270EA
101.85
USD
76910-270
76910-272
76910-274
4-Bromo-2-(trifluoromethyl)benzeneboronic acid ≥95%
4-Bromo-2-(trifluoromethyl)benzeneboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BBrF₃O₂ MW: 268.824 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18447612 CAS Number: 1394346-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...