3-(Cyclopropylmethoxy)-2-fluoro-pyridine ≥97%

Supplier: AOBChem USA

Warning

20643-1G 20643-25G 20643-5G
76910-260EA 79.57 USD
76910-260 76910-262 76910-264
3-(Cyclopropylmethoxy)-2-fluoro-pyridine ≥97%
3-(Cyclopropylmethoxy)-2-fluoro-pyridine
Formula: C₉H₁₀FNO
MW: 167.18 g/mol
Storage Temperature: Ambient
CAS Number: 205594-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR