3-Ethoxy-4-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20565-1G 20565-25G 20565-500MG 20565-5G
76910-236EA 126.25 USD
76910-236 76910-238 76910-240 76910-242
3-Ethoxy-4-fluorobenzaldehyde ≥97%
3-Ethoxy-4-fluorobenzaldehyde
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12022599
CAS Number: 870837-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR