3-Chloro-4-isopropoxyphenol ≥97%

Supplier: AOBChem USA

Warning

20448-1G 20448-25G 20448-500MG 20448-5G
76910-186EA 165.51 USD
76910-186 76910-188 76910-190 76910-192
3-Chloro-4-isopropoxyphenol ≥97%
3-Chloro-4-isopropoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MW: 186.635 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14537050
CAS Number: 1216247-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR