3,6-Dibromo-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

20439-1G 20439-25G 20439-500MG 20439-5G
76910-172EA 260.99 USD
76910-172 76910-174 76910-176 76910-178
3,6-Dibromo-2-fluorophenol ≥97%
3,6-Dibromo-2-fluorophenol
Formula: C₆H₃Br₂FO
MW: 269.894 g/mol
Storage Temperature: Ambient
CAS Number: 1803817-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR